Learn More
Cayman Chemical ANTIBODY TAS 120 5mg
SDP
Supplier:  Cayman Chemical 211365 MG
Encompass_Preferred
An orally bioavailable inhibitor of FGFRs that irreversibly inhibits all four FGFR subtypes at nanomolar concentrations; blocks the proliferation of human cancer cell lines with FGFR gene abnormalities
Catalog No. 50-298-1147