Learn More
Cayman Chemical ANTIBODY TBA-354 250mg
SDP
Supplier:  Cayman Chemical 30317250 MG
Encompass_Preferred
An antitubercular agent; active against clinical isolates of drug-resistant and -sensitive replicating M. tuberculosis (MICs = M. tuberculosis over M. scrofulaceum, M. gilvum, M. fortuitum, M. triviale, and M. smegmatis (MICs = >11.5 μM) but is active against M. bovis and M. kansasii (MICs = M. tuberculosis infection at 100 mg/kg per day
Catalog No. 50-299-2033