Learn More
Cayman Chemical ANTIBODY TCID 10mg
SDP
Supplier:  Cayman Chemical 1635310 MG
Encompass_Preferred
A tetrachloro indandione that acts as a potent inhibitor of UCH-L3 and demonstrates 125-fold selectivity over UCH-L1 (IC50s = 0.6 and 75 µM, respectively)
Catalog No. 50-297-3059