Learn More
Cayman Chemical ANTIBODY TCN 201 1mg
SDP
Supplier:  Cayman Chemical 190861 MG
Encompass_Preferred
A selective antagonist of NMDA receptors containing the NR2A subunit (pIC50s = 6.8 and <4.3 for human recombinant NR2A and NR2B, respectively); binds to a novel allosteric site located at the dimer interface between the GluN1 and GluN2 binding domains, thereby reducing glycine signal transduction and inhibiting NMDA receptor function
Catalog No. 50-298-1599