Learn More
Cayman Chemical ANTIBODY TCS 1102 25mg
SDP
Supplier:  Cayman Chemical 1849525 MG
Encompass_Preferred
A dual antagonist of both OX1R and OX2R (Kis = 0.2 and 3 nM, respectively) that demonstrates good brain penetration when administered intraperitoneally; promotes sleep, prevents drug-induced plasticity and drug relapse, and decreases fear and anxiety in response to acute episodes of stress
Catalog No. 50-297-9953