Learn More
Cayman Chemical ANTIBODY Tegoprazn 10mg
SDP
Supplier:  Cayman Chemical 3324310 MG
Encompass_Preferred
A proton pump inhibitor (IC50 = 0.53 µM in porcine gastric membrane vesicles); inhibits basal gastric acid secretion in pylorus-ligated rats at 10 mg/kg; reduces esophageal mucosal tissue MPO levels in a rat model of reflux esophagitis (ED50 = 5.9 mg/kg); reduces ulcer area in rat models of naproxen-, WIRS-, or ethanol-induced gastric ulcers (ED50s = 0.1, 0.1, and 1.4 mg/kg, respectively)
Catalog No. 50-294-7009