Learn More
Cayman Chemical ANTIBODY TEMPOn 10g
SDP
Supplier:  Cayman Chemical 1487710 G
Encompass_Preferred
The 4-oxo derivative of TEMPO; possible uses as a spin trap, in hydrogen transfer experiments, and as a polarizing agent in dynamic nuclear polarization NMR spectroscopy
Catalog No. 50-296-8688