Learn More
Cayman Chemical ANTIBODY TenovIn-3 500ug
SDP
Supplier:  Cayman Chemical 16288500 UG
Encompass_Preferred
A small molecule activator of p53; increases p53 activity in MCF-7 cells at 10 µM; increases the level of K40-acetylated α-tubulin in H1299 cells; inhibits SIRT2 protein deacetylase activity
Catalog No. 50-297-2623