Learn More
Cayman Chemical ANTIBODY TFAP 25mg
SDP
Supplier:  Cayman Chemical 2674525 MG
Encompass_Preferred
An inhibitor of COX-1 (IC50 = 0.8 μM); selective for COX-1 over COX-2 (IC50 = 210 μM); reduces acetic acid-induced writhing in mice at 10 and 30 mg/kg; has analgesic activity in the formalin test in rats at 30 mg/kg; does not induce gastric damage in rats at doses up to 300 mg/kg
Catalog No. 50-293-6950