Learn More
Cayman Chemical ANTIBODY TH-302 25mg
SDP
Supplier:  Cayman Chemical 2219525 MG
Encompass_Preferred
A hypoxia-activated prodrug and DNA alkylating agent with anticancer activity; selectively cytotoxic to H460 human NSCLC cells grown under hypoxic over normoxic conditions (IC50s = 0.019 and 5.1 μM, respectively); reduces survival of H460 and HT-29 cells in a clonogenic assay (IC50s = 0.2 and 0.2 μM, respectively); inhibits primary tumor growth by 41% in an MIA PaCa-2-RFP mouse orthotopic xenograft model at 33 mg/kg per day; inhibits tumor growth in the Calu-6 NSCLC, H82 small cell lung, A375 melanoma, PC-3 prostate, and BxPC-3 pancreatic cancer mouse xenograft models at 50 mg/kg
Catalog No. 50-298-6343