Learn More
Cayman Chemical ANTIBODY TH287 10mg
SDP
Supplier:  Cayman Chemical 1813210 MG
Encompass_Preferred
A selective inhibitor of the nucleotide pool sanitizing enzyme MTH1 (IC50 = 0.8 nM) that kills cancer cell lines (IC50s = 0.8-3.06 µM)
Catalog No. 50-296-5102