Learn More
Cayman Chemical ANTIBODY ThIamet G 10mg
SDP
Supplier:  Cayman Chemical 1323710 MG
Encompass_Preferred
A potent and selective inhibitor of O-GlcNAcase (Ki = 21 nM); increases cellular O-GlcNAc-modified protein levels (EC50 = 30 nM) and blocks phosphorylation of tau protein both in cultured PC-12 cells and in rats (200 mg/kg/day); is orally bioavailable and effectively cross the blood brain barrier
Catalog No. 50-295-2988