Learn More
Cayman Chemical ANTIBODY ThIoLox 10mg
SDP
Supplier:  Cayman Chemical 3894610 MG
Encompass_Preferred
A 15-LO-1 inhibitor (Ki = 3.3 µM); a P. falciparum aspartate carbamoyltransferase inhibitor (IC50 = 3.32 µM); reduces LPS-induced expression of the mRNAs encoding TNF-α, iNos, Il-1β, Il-6, Il-8, and Il-12b in mouse lung slices at 50 µM; inhibits glutamate-induced increases in lipid peroxidation and mitochondrial superoxide formation, as well as cell death, in mouse HT22 cells at 10 µM
Catalog No. 50-291-7766