Learn More
Cayman Chemical ANTIBODY TIC10 10mg
SDP
Supplier:  Cayman Chemical 1610910 MG
Encompass_Preferred
Induces the expression of TRAIL at 1-5 µM in a p53-independent manner; orally active, stable, and crosses the blood-brain barrier; suppresses the growth of orthotopic human glioblastoma multiforme tumors in mice
Catalog No. 50-297-1636