Learn More
Cayman Chemical ANTIBODY TIcagrelor 25mg
SDP
Supplier:  Cayman Chemical 1542525 MG
Encompass_Preferred
A reversible purinergic P2Y12 receptor antagonist (Ki = 14 nM; IC50 = 1.8 μM)
Catalog No. 50-297-2655