Learn More
Cayman Chemical ANTIBODY TIoxolon 5g
SDP
Supplier:  Cayman Chemical 313725 G
Encompass_Preferred
A carbonic anhydrase I inhibitor (Ki = 91 nM); selective for CAI over CAII, -III, -IV, -VA, -VB, -VI, -VII, -IX, -XII, and -XIV (Kis = 4,930-9,040 nM); active against various Gram-positive or -negative bacteria, including M. tuberculosis; active against dermatophytes, yeasts, and molds
Catalog No. 50-299-1884