Learn More
Cayman Chemical ANTIBODY TobramycIn 500mg
SDP
Supplier:  Cayman Chemical 14596500 MG
Encompass_Preferred
An aminoglycoside antibiotic; binds to the 30S ribosomal subunit, inhibiting protein synthesis; active against a variety of Gram-positive and Gram-negative bacteria; reduces the number of CFUs in several mouse models of thigh infection
Catalog No. 50-297-2097