Learn More
Cayman Chemical ANTIBODY Tozadent 100mg
SDP
Supplier:  Cayman Chemical 28452100 MG
Encompass_Preferred
An adenosine A2A receptor antagonist (Kis = 11.5 and 6 nM for human and rhesus monkey receptors, respectively); increases the distance traveled and reduces contralateral asymmetry in the open field test in a rat model of Parkinson's disease induced by 6-OHDA at 30 mg/kg; reverses locomotor deficits and restores novel object-stimulated locomotion in a marmoset model of MPTP-induced Parkinson's disease at 150 mg/kg
Catalog No. 50-294-1794