Learn More
Cayman Chemical ANTIBODY TP-0463518 10mg
SDP
Supplier:  Cayman Chemical 3378810 MG
Encompass_Preferred
A pan HIF-PH inhibitor; inhibits human HIF-PH1 (Ki = 5.3 nM), as well as human HIF-PH2 and HIF-PH3 (IC50s = 18 and 63 nM, respectively); increases serum levels of EPO in mice at 5 mg/kg; increases serum EPO levels in a 5/6 nephrectomy-induced rat model of chronic kidney disease at 10 mg/kg
Catalog No. 50-294-7461