Learn More
Cayman Chemical ANTIBODY TPX-0005 5mg
SDP
Supplier:  Cayman Chemical 233975 MG
Encompass_Preferred
A multi-kinase inhibitor; inhibits TrkA, TrkB, and TrkC (IC50s = 0.83, 0.05, and 0.1 nM, respectively) as well as ROS1 (IC50 = 0.07 nM); inhibits a variety of other kinases, including ALK, JAK2, LYN, Src, and FAK (IC50s = 1.04, 1.04, 1.66, 5.3, and 6.96 nM, respectively); inhibits proliferation of PC-9, H1975, 11-18, HCC4006, and HCC827 cells (IC50s = 100, 100, 150, 148, and 430 nM, respectively) with additive or synergistic effects when used in combination with certain compounds, including osimertinib; reduces tumor growth in PC-9 and H1975 mouse xenograft models; potentiates the effect of osimertinib on tumor growth in vivo
Catalog No. 50-296-3924