Learn More
Cayman Chemical ANTIBODY trasemIde 10mg
SDP
Supplier:  Cayman Chemical 2131210 MG
Encompass_Preferred
An analytical reference standard categorized as a diuretic; has been abused as a performance-enhancing drug and masking agent in doping in sports; intended for research and forensic applications
Catalog No. 50-298-5337