Learn More
Cayman Chemical ANTIBODY TSI-01 10mg
SDP
Supplier:  Cayman Chemical 1762810 MG
Encompass_Preferred
A selective inhibitor of LPCAT2 (IC50s = 0.47 versus 3.02 µM for human LPCAT2 and LPCAT1, respectively) that can suppress PAF biosynthesis in mouse peritoneal macrophages stimulated with a calcium ionophore
Catalog No. 50-298-0173