Learn More
Cayman Chemical ANTIBODY TX1-85-1 10mg
SDP
Supplier:  Cayman Chemical 1916410 MG
Encompass_Preferred
An inhibitor of ErbB3 (IC50 = 23 nM); binds to ErbB2, Lyn, and several other Src family kinases at 2 µM; inhibits phosphorylation of Akt and ERK1/2 in ErbB3-dependent HCC2935 and OVCAR-8 cells at 5 and 2 µM, respectively; does not inhibit proliferation of ErbB3-dependent PC9 GR4, HCC827 GR8, or OVCAR-8 cells (EC50s = ≥10 µM)
Catalog No. 50-298-1756