Learn More
Cayman Chemical ANTIBODY TZ9 10mg
SDP
Supplier:  Cayman Chemical 1596410 MG
Encompass_Preferred
A cell-permeable triazine compound that inhibits the human E2 ubiquitin-conjugating enzyme Rad6B; selectively docks to the Rad6B catalytic site, inhibits Rad6B-induced histone H2A ubiquitination, downregulates intracellular β-catenin, induces G2/M arrest and apoptosis, and inhibits proliferation and migration of metastatic human breast cancer cells
Catalog No. 50-297-0372