Learn More
Cayman Chemical ANTIBODY U-0126 10mg
SDP
Supplier:  Cayman Chemical 7097010 MG
Encompass_Preferred
A non ATP-competitive MEK inhibitor (IC50 = 72 and 58 nM for MEK1 and MEK2, respectively); an activator of AMPK (EC50 = 15 µM); increases the ratios of ADP:ATP and AMP:ATP
Catalog No. 50-289-6495