Learn More
Cayman Chemical ANTIBODY U-74389G 100mg
SDP
Supplier:  Cayman Chemical 75860100 MG
Encompass_Preferred
U-74389G is an antioxidant which prevents iron-dependent lipid peroxidation. It protects against is chemia-reperfusion injury in animal heart, liver, and kidney models.{4561,4612,4562,4567}
Catalog No. 50-289-4827