Learn More
Cayman Chemical ANTIBODY UCF 101 25mg
SDP
Supplier:  Cayman Chemical 1555025 MG
Encompass_Preferred
A specific inhibitor of the proteolytic activity of Omi/HtrA2 (IC50 = 9.5 μM) that is naturally fluorescent, thus enabling the visualization of its entry into cells; has been reported to decrease cerebral infarct size in a rat model of cerebral ischemia and to be protective of cardiomyocytes against ischemic injury
Catalog No. 50-296-9333