Learn More
Cayman Chemical ANTIBODY UNBS5162 5mg
SDP
Supplier:  Cayman Chemical 177395 MG
Encompass_Preferred
An antagonist of CXCL chemokine expression; demonstrates antiangiogenic properties both in vitro and in vivo in prostate cancer research models
Catalog No. 50-296-7639