Learn More
Cayman Chemical ANTIBODY UNC0379 25mg
SDP
Supplier:  Cayman Chemical 1640025 MG
Encompass_Preferred
A selective inhibitor of the lysine methyltransferase SET8 (IC50 = 7.3 µM; Kd = 18.3 µM)
Catalog No. 50-297-0978