Learn More
Cayman Chemical ANTIBODY UNC2025 25mg
SDP
Supplier:  Cayman Chemical 1661325 MG
Encompass_Preferred
A potent, orally bioavailable inhibitor of the tyrosine kinases Mer and Flt3 (IC50s = 0.74 and 0.80 nM, respectively); blocks colony formation of Mer- and Flt3-dependent tumor cell lines and inhibits Mer phosphorylation in bone marrow leukemia cells in vivo
Catalog No. 50-296-7332