Learn More
Cayman Chemical ANTIBODY UNC2250 10mg
SDP
Supplier:  Cayman Chemical 1770310 MG
Encompass_Preferred
A selective inhibitor of Mer kinase activity, blocking the steady-state phosphorylation of endogenous Mer in 697 B-ALL cells with an IC50 value of 9.8 nM; inhibits Mer activity in cells and displays good pharmacokinetic properties in mice following intravenous or oral administration
Catalog No. 50-296-8070