Learn More
Cayman Chemical ANTIBODY UPF-1069 1mg
SDP
Supplier:  Cayman Chemical 177601 MG
Encompass_Preferred
A PARP inhibitor that demonstrates a modest preference for PARP2 over PARP1 (IC50s = 0.3 and 8 µM, respectively)
Catalog No. 50-296-7642