Learn More
Cayman Chemical ANTIBODY UprosertIb 25mg
SDP
Supplier:  Cayman Chemical 1990425 MG
Encompass_Preferred
A selective, orally bioavailable inhibitor of Akt (IC50s = 180, 328, and 38 nM for Akt1, Akt2, and Akt3, respectively); preferentially inhibits the proliferation of human cancer cells lines with Akt pathway activation via PI3K/PTEN mutation or loss
Catalog No. 50-298-0668