Learn More
Cayman Chemical ANTIBODY UrIdIn 25g
SDP
Supplier:  Cayman Chemical 2030025 G
Encompass_Preferred
A pyrimidine nucleoside; forms complementary base pairs with adenosine in RNA; replaced with thymidine in DNA
Catalog No. 50-298-1188