Learn More
Cayman Chemical ANTIBODY UsnIc AcId 5g
SDP
Supplier:  Cayman Chemical 225855 G
Encompass_Preferred
A lichen metabolite with antioxidant, anticancer, antifungal, antiprotozoal, and antimicrobial activities; reduces the number of indomethacin-induced gastric ulcers in rats through decreased lipid peroxidation and decreased myeloperoxidase activity; fungicidal against C. orthopsilosis and C. parapsilosis (IC80s = 7.8 and 15.6 μg/ml, respectively); bactericidal against vancomycin-resistant E. faecalis (MIC = 125 μg/ml); reduces L. amazonensis parasite loads by 72.3% in footpads of infected mice; reduces growth of cancer cells in vitro (IC50s = 48.5-199 μM)
Catalog No. 50-298-6531