Learn More
Cayman Chemical ANTIBODY USP8-IN-3 5mg
SDP
Supplier:  Cayman Chemical 416005 MG
Encompass_Preferred
A USP8 inhibitor (IC50 = 4 µM); inhibits the growth of H1975 and GH3 cells (GI50s = 6.01 and 37.03 µM)
Catalog No. 50-294-3197