Learn More
Cayman Chemical ANTIBODY UTL-5g 50mg
SDP
Supplier:  Cayman Chemical 4224250 MG
Encompass_Preferred
A chemo- and radioprotective agent; reduces LPS-induced transcriptional activity mediated by a variety of immune-related transcription factors, including STAT1, STAT2, STAT3, and SMAD2-4 but not NF-κB in RAW 264.7 macrophages; reduces radiation-induced increases in liver TNF-α levels in mice at 30 and 60 mg/kg; protective against liver radiation-induced hepatotoxicity in mice at 60 mg/kg; increases the survival rate in a mouse model of acute cisplatin toxicity
Catalog No. 50-294-3540