Learn More
Cayman Chemical ANTIBODY ValrubIcIn 5mg
SDP
Supplier:  Cayman Chemical 229645 MG
Encompass_Preferred
A semisynthetic anthracycline antitumor antibiotic; inhibits the growth of human bladder tumor cell lines (LD50s = 0.05-11.2 μM); more active against cells in the plateau growth phase than in the log growth phase (LD70s = 2.5 and 12.1 μM, respectively for CUB-2 cells); increases survival in mice with murine P388 and L1210 leukemias; reduces epidermal thickness and normalizes epidermal morphology in a mouse psoriasis xenograft transplantation model
Catalog No. 50-299-4475