Learn More
Cayman Chemical ANTIBODY Velaresol 5mg
SDP
Supplier:  Cayman Chemical 371905 MG
Encompass_Preferred
An Hb-O2 stabilizer; increases erythrocyte deformability in isolated human blood from a patient with sickle cell disease at 1.5 mM; inhibits deoxygenation-induced decreases in erythrocyte deformability in isolated human blood from a patient with sickle cell disease at 3 or 5 mM; decreases oxygen tension and increases oxygen saturation of isolated human sickle erythrocytes at 5 mM
Catalog No. 50-291-5136