Learn More
Cayman Chemical ANTIBODY VIdarabIn 1g
SDP
Supplier:  Cayman Chemical 181491 G
Encompass_Preferred
An analog of the nucleoside adenosine that has antiviral properties; acts as a prodrug that, once phosphorylated by cellular enzymes, acts as both substrate and inhibitor of DNA polymerase
Catalog No. 50-296-7677