Learn More
Cayman Chemical ANTIBODY vIFSP1 500ug
SDP
Supplier:  Cayman Chemical 39927500 UG
Encompass_Preferred
An inhibitor of FSP1 (IC50s = 34 and 83 nM for the human and mouse enzymes, respectively); induces ferroptosis in Gpx4 knockout Pfa1 mouse embryonic fibroblasts expressing human or mouse FSP1, an effect that can be reversed by liproxstatin-1
Catalog No. 50-294-5053