Learn More
Cayman Chemical ANTIBODY VR23 5mg
SDP
Supplier:  Cayman Chemical 191335 MG
Encompass_Preferred
A trypsin-like proteasome inhibitor (IC50 = 1 nM) that deregulates the activity of cyclin E and other centrosomal proteins, resulting in the induction of multiple centrosome amplification, abnormal spindle formation, uneven cytokinesis, irreversible mitotic arrest, and eventually apoptosis that is specific to cancer cells
Catalog No. 50-298-1098