Learn More
Cayman Chemical ANTIBODY VT103 1mg
SDP
Supplier:  Cayman Chemical 423271 MG
Encompass_Preferred
A TEAD1 inhibitor (IC50 = 1.02 nM); inhibits TEAD autopalmitoylation in HEK293 cells expressing TEAD1 but not HEK293 cells expressing TEAD2, TEAD3, or TEAD4 at 3 µM; inhibits palmitoylation of endogenous TEAD1 in NCI-H2373 cells; reduces tumor volume in NCI-H2373 and NCI H226 mesothelioma mouse xenograft models
Catalog No. 50-297-8500