Learn More
Cayman Chemical ANTIBODY VU0155069 10mg
SDP
Supplier:  Cayman Chemical 1320610 MG
Encompass_Preferred
A potent and selective inhibitor of PLD1, both in vitro (IC50 = 46 nM) and in cells (IC50 = 11 nM); also effective as a PLD2 inhibitor at higher concentrations (IC50 = 933 nM in vitro; 1,800 nM in cells); strongly inhibits the invasive migration of several breast cancer cell lines in trans well assays
Catalog No. 50-295-2298