Learn More
Cayman Chemical ANTIBODY VU0359595 25mg
SDP
Supplier:  Cayman Chemical 1095525 MG
Encompass_Preferred
A highly selective inhibitor of PLD1 (IC50 = 3.7 nM) versus PLD2 (IC50 = 6.4 μM)
Catalog No. 50-295-2505