Learn More
Cayman Chemical ANTIBODY Wnt-C59 5mg
SDP
Supplier:  Cayman Chemical 166445 MG
Encompass_Preferred
A potent inhibitor of PORCN (IC50 = 74 pM), completely blocking Wnt secretion into culture media; prevents activation of all evaluated Wnt family members; effective in vivo as well as in vitro; blocks the progression of breast cancer while downregulating Wnt/β-catenin pathway target genes in mice
Catalog No. 50-296-6987