Learn More
Cayman Chemical ANTIBODY WP1130 50mg
SDP
Supplier:  Cayman Chemical 1522750 MG
Encompass_Preferred
Inhibits the deubiquitinase activity of USP9x, USP5, USP14, and UCH37; 5 μM induces the accumulation of protein-ubiquitin conjugates, resulting in the formation of aggresomes and apoptosis in a variety of tumor cells
Catalog No. 50-296-9214