Learn More
Cayman Chemical ANTIBODY WWL113 25mg
SDP
Supplier:  Cayman Chemical 1618125 MG
Encompass_Preferred
A selective inhibitor of the mouse carboxylesterases Ces3 and Ces 1f (IC50 = ~0.1 µM); corrects multiple features of metabolic syndrome in obese-diabetic db/db mice and in mice with diet-induced obesity; also inhibits hCES1, the human ortholog of mouse Ces3 (IC50 = ~50 nM)
Catalog No. 50-296-8630