Learn More
Cayman Chemical ANTIBODY WWL123 1mg
SDP
Supplier:  Cayman Chemical 168491 MG
Encompass_Preferred
A brain-penetrant, selective inhibitor of ABHD6 (IC50 = 0.43 µM) that has been used to decrease seizure incidence in a genetic mouse model of juvenile Huntington’s disease as well as in chemically-induced epilepsy models
Catalog No. 50-296-7034