Learn More
Cayman Chemical ANTIBODY WYnC 25mg
SDP
Supplier:  Cayman Chemical 4200925 MG
Encompass_Preferred
A chemoselective probe for protein sulfenylation; selective for a sulfenylated dipeptide over sulfinic acid- or sulfonic acid-containing dipeptides, reduced GSH, GSSG, and GSNO, as well as cysteine sulfinic acid and glutathione sulfonic acid in cell-free assays; selectively binds to C64,82S GPX3 containing a sulfenic acid at Cys36 over C64,82S GPX3 containing a reduced thiol, sulfinic acid at Cys36, or an intramolecular disulfide bond in cell-free assays; binds to sulfenylated proteins in live cells and can then be conjugated to azide-containing fluorescent probes for detection
Catalog No. 50-297-8495